Assay Details
Gene Symbol: mapk14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131722.1 | NP_571797.1 | 10 - 11 | 1089 | 89 | |
XM_005167061.4 | XP_005167118.1 | 10 - 11 | 1220 | 89 | ||
GenBank mRNA | AB030897.1 | - | 10 - 11 | 1089 | 89 | |
BC044128.1 | - | 10 - 11 | 924 | 89 | ||
BC059417.1 | - | 10 - 11 | 1094 | 89 | ||
BC164402.1 | - | 10 - 11 | 883 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
