Assay Details
Gene Symbol: gstp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131734.3 | NP_571809.1 | 2 - 3 | 111 | 66 | |
GenBank mRNA | AB194127.1 | - | 2 - 3 | 66 | 66 | |
AF285098.1 | - | 2 - 3 | 66 | 66 | ||
BC162358.1 | - | 2 - 3 | 166 | 66 | ||
BC162376.1 | - | 2 - 3 | 166 | 66 | ||
CK361426.1 | - | 2 - 3 | 151 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
