Assay Details
| Gene Symbol: gnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131869.2 | NP_571944.1 | 8 - 9 | 1263 | 93 | |
| GenBank mRNA | BC059498.1 | - | 8 - 9 | 1263 | 93 | |
More Information
Set Membership: |
|

gnat2
Genomic Map