Assay Details
Gene Symbol: epha4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153658.1 | NP_705944.1 | 2 - 3 | 197 | 158 | |
GenBank mRNA | AF369381.1 | - | 2 - 3 | 197 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
