Assay Details
| Gene Symbol: selenow1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178287.5 | NP_840072.3 | 2 - 3 | 191 | 69 | |
| GenBank mRNA | AY216582.1 | - | 2 - 3 | 102 | 69 | |
| BC092686.1 | - | 2 - 3 | 153 | 69 | ||
| BC152097.1 | - | 2 - 3 | 127 | 69 | ||
| CK395720.1 | - | 2 - 3 | 202 | 69 | ||
| DN892184.1 | - | 2 - 3 | 172 | 69 | ||
| EH445855.1 | - | 2 - 3 | 214 | 69 | ||
| EH564833.1 | - | 4 - 5 | 651 | 69 | ||
| EH593497.1 | - | 2 - 3 | 224 | 69 | ||
More Information
Set Membership: |
|

selenow1
Genomic Map