Assay Details
| Gene Symbol: cxcl12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178307.2 | NP_840092.1 | 3 - 4 | 330 | 65 | |
| GenBank mRNA | AY147915.1 | - | 3 - 4 | 264 | 65 | |
| AY577011.1 | - | 3 - 4 | 330 | 65 | ||
| BC092687.1 | - | 3 - 4 | 316 | 65 | ||
| BC109417.1 | - | 3 - 4 | 325 | 65 | ||
| FJ915063.1 | - | 3 - 4 | 294 | 65 | ||
More Information
Set Membership: |
|

cxcl12a
Genomic Map