Assay Details
Gene Symbol: tmpoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180968.3 | NP_851299.1 | 12 - 13 | 1441 | 68 | |
XM_005164721.4 | XP_005164778.1 | 9 - 10 | 1147 | 68 | ||
GenBank mRNA | AJ320190.1 | - | 12 - 13 | 1389 | 68 | |
BC048056.1 | - | 12 - 13 | 1441 | 68 | ||
BC165860.1 | - | 12 - 13 | 1417 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
