Assay Details
Gene Symbol: tmpoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328563.1 | NP_001315492.1 | 7 - 8 | 1000 | 72 | |
NM_180968.3 | NP_851299.1 | 7 - 8 | 1000 | 72 | ||
XM_005164720.4 | XP_005164777.1 | 7 - 8 | 1007 | 72 | ||
GenBank mRNA | AJ320189.1 | - | 7 - 8 | 948 | 72 | |
BC048056.1 | - | 7 - 8 | 1000 | 72 | ||
BC165860.1 | - | 7 - 8 | 976 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
