Assay Details
Gene Symbol: tmpoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328563.1 | NP_001315492.1 | 8 - 9 | 1065 | 70 | |
NM_180968.3 | NP_851299.1 | 8 - 9 | 1065 | 70 | ||
XM_005164720.4 | XP_005164777.1 | 8 - 9 | 1072 | 70 | ||
XM_005164721.4 | XP_005164778.1 | 5 - 6 | 771 | 70 | ||
XM_005164722.4 | XP_005164779.1 | 5 - 6 | 771 | 70 | ||
GenBank mRNA | AJ320189.1 | - | 8 - 9 | 1013 | 70 | |
AJ320190.1 | - | 8 - 9 | 1013 | 70 | ||
AJ320191.1 | - | 5 - 6 | 713 | 70 | ||
BC048056.1 | - | 8 - 9 | 1065 | 70 | ||
BC165860.1 | - | 8 - 9 | 1041 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
