Assay Details
| Gene Symbol: aldocb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194384.1 | NP_919365.1 | 5 - 6 | 610 | 62 | |
| GenBank mRNA | AF533647.1 | - | 5 - 6 | 616 | 62 | |
| BC053192.1 | - | 5 - 6 | 610 | 62 | ||
| BC164189.1 | - | 4 - 5 | 577 | 62 | ||
More Information
Set Membership: |
|

aldocb
Genomic Map