Assay Details
| Gene Symbol: aldocb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194384.1 | NP_919365.1 | 6 - 7 | 693 | 60 | |
| GenBank mRNA | AF533647.1 | - | 6 - 7 | 699 | 60 | |
| BC053192.1 | - | 6 - 7 | 693 | 60 | ||
| BC164189.1 | - | 5 - 6 | 660 | 60 | ||
More Information
Set Membership: |
|

aldocb
Genomic Map