Assay Details
| Gene Symbol: agt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198063.1 | NP_932329.1 | 2 - 3 | 822 | 89 | |
| GenBank mRNA | AY049731.1 | - | 2 - 3 | 822 | 89 | |
| BC095585.1 | - | 2 - 3 | 832 | 89 | ||
More Information
Set Membership: |
|

agt
Genomic Map