Assay Details
| Gene Symbol: tpd52l2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199582.2 | NP_955876.2 | 2 - 3 | 359 | 82 | |
| NM_200885.2 | NP_957179.1 | 2 - 3 | 359 | 82 | ||
| XM_005162032.2 | XP_005162089.1 | 2 - 3 | 374 | 82 | ||
| XM_005162033.3 | XP_005162090.1 | 2 - 3 | 374 | 82 | ||
| XM_005162035.2 | XP_005162092.1 | 2 - 3 | 374 | 82 | ||
| XM_005162036.3 | XP_005162093.1 | 2 - 3 | 374 | 82 | ||
| XM_005162037.2 | XP_005162094.1 | 2 - 3 | 374 | 82 | ||
| XM_021469740.1 | XP_021325415.1 | 2 - 3 | 374 | 82 | ||
| XM_021469741.1 | XP_021325416.1 | 2 - 3 | 374 | 82 | ||
| XM_021469742.1 | XP_021325417.1 | 2 - 3 | 374 | 82 | ||
| XM_021469744.1 | XP_021325419.1 | 2 - 3 | 374 | 82 | ||
| XM_021469745.1 | XP_021325420.1 | 2 - 3 | 374 | 82 | ||
| GenBank mRNA | BC045297.1 | - | 2 - 3 | 339 | 82 | |
| BC063969.1 | - | 2 - 3 | 325 | 82 | ||
| EH444392.1 | - | 2 - 3 | 368 | 82 | ||
More Information
Set Membership: |
|

tpd52l2b
Genomic Map