Assay Details
| Gene Symbol: tpd52l2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199582.2 | NP_955876.2 | 6 - 7 | 715 | 89 | |
| NM_200885.2 | NP_957179.1 | 5 - 6 | 667 | 89 | ||
| XM_005162032.2 | XP_005162089.1 | 8 - 9 | 802 | 89 | ||
| XM_005162033.3 | XP_005162090.1 | 8 - 9 | 799 | 89 | ||
| XM_005162035.2 | XP_005162092.1 | 7 - 8 | 754 | 89 | ||
| XM_005162036.3 | XP_005162093.1 | 7 - 8 | 751 | 89 | ||
| XM_005162037.2 | XP_005162094.1 | 6 - 7 | 724 | 89 | ||
| XM_021469740.1 | XP_021325415.1 | 9 - 10 | 862 | 89 | ||
| XM_021469741.1 | XP_021325416.1 | 9 - 10 | 859 | 89 | ||
| XM_021469742.1 | XP_021325417.1 | 8 - 9 | 832 | 89 | ||
| XM_021469744.1 | XP_021325419.1 | 7 - 8 | 790 | 89 | ||
| GenBank mRNA | BC045297.1 | - | 6 - 7 | 695 | 89 | |
| BC063969.1 | - | 5 - 6 | 633 | 89 | ||
| EH444392.1 | - | 6 - 7 | 724 | 89 | ||
More Information
Set Membership: |
|

tpd52l2b
Genomic Map