Assay Details
| Gene Symbol: bzw1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199708.1 | NP_956002.1 | 2 - 3 | 164 | 111 | |
| GenBank mRNA | BC058875.1 | - | 2 - 3 | 164 | 111 | |
More Information
Set Membership: |
|

bzw1a
Genomic Map