Assay Details
| Gene Symbol: ier5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199754.1 | NP_956048.1 | 1 | 946 | 89 | |
| GenBank mRNA | BC059641.1 | - | 1 | 946 | 89 | |
| BC066470.1 | - | 1 | 954 | 89 | ||
More Information
Set Membership: |
|

ier5l
Genomic Map