Assay Details
| Gene Symbol: ppp2r2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309438.1 | NP_001296367.1 | 5 - 6 | 748 | 97 | |
| GenBank mRNA | BC056824.1 | - | 4 - 5 | 431 | 97 | |
More Information
Set Membership: |
|

ppp2r2ab
Genomic Map