Assay Details
Gene Symbol: ppp2r2ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309438.1 | NP_001296367.1 | 9 - 10 | 1376 | 76 | |
GenBank mRNA | BC056824.1 | - | 8 - 9 | 1059 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
