Assay Details
| Gene Symbol: ppp2r2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309438.1 | NP_001296367.1 | 10 - 11 | 1464 | 82 | |
| GenBank mRNA | BC056824.1 | - | 9 - 10 | 1147 | 82 | |
More Information
Set Membership: |
|

ppp2r2ab
Genomic Map