Assay Details
Gene Symbol: utp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199998.1 | NP_956292.1 | 7 - 8 | 790 | 122 | |
GenBank mRNA | BC049053.1 | - | 7 - 8 | 790 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
