Assay Details
Gene Symbol: imp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291357.1 | NP_001278286.1 | 1 - 2 | 231 | 146 | |
GenBank mRNA | EH493453.1 | - | 1 - 2 | 296 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
