Assay Details
Gene Symbol: imp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291357.1 | NP_001278286.1 | 4 - 5 | 495 | 68 | |
GenBank mRNA | EH466588.1 | - | 3 - 4 | 410 | 68 | |
EH493453.1 | - | 4 - 5 | 560 | 68 | ||
EH598772.1 | - | 3 - 4 | 440 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
