Assay Details
| Gene Symbol: rnf175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328343.1 | NP_001315272.1 | 5 - 6 | 548 | 82 | |
| NM_200308.3 | NP_956602.2 | 6 - 7 | 585 | 82 | ||
| XM_021466728.1 | XP_021322403.1 | 5 - 6 | 564 | 82 | ||
| GenBank mRNA | BC050174.1 | - | 6 - 7 | 586 | 82 | |
| BC071470.1 | - | 5 - 6 | 539 | 82 | ||
More Information
Set Membership: |
|

rnf175
Genomic Map