Assay Details
| Gene Symbol: eef2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200458.2 | NP_956752.2 | 2 - 3 | 272 | 59 | |
| GenBank mRNA | AY391422.1 | - | 2 - 3 | 272 | 59 | |
| BC045488.1 | - | 2 - 3 | 303 | 59 | ||
| BC063965.1 | - | 2 - 3 | 283 | 59 | ||
More Information
Set Membership: |
|

eef2b
Genomic Map