Assay Details
| Gene Symbol: phactr4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200670.1 | NP_956964.1 | 14 - 15 | 2473 | 79 | |
| XM_005158332.4 | XP_005158389.1 | 13 - 14 | 2858 | 79 | ||
| XM_005158333.4 | XP_005158390.1 | 13 - 14 | 3086 | 79 | ||
| XM_005158334.4 | XP_005158391.1 | 15 - 16 | 2852 | 79 | ||
| XM_005158335.4 | XP_005158392.1 | 12 - 13 | 2724 | 79 | ||
| XM_009292432.3 | XP_009290707.1 | 15 - 16 | 3352 | 79 | ||
| XM_009292434.3 | XP_009290709.1 | 14 - 15 | 2720 | 79 | ||
| XM_021466650.1 | XP_021322325.1 | 15 - 16 | 2800 | 79 | ||
| GenBank mRNA | BC058052.1 | - | 14 - 15 | 2473 | 79 | |
More Information
Set Membership: |
|

phactr4b
Genomic Map