Assay Details
| Gene Symbol: phactr4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200670.1 | NP_956964.1 | 5 - 6 | 485 | 85 | |
| XM_005158332.4 | XP_005158389.1 | 3 - 4 | 570 | 85 | ||
| XM_005158333.4 | XP_005158390.1 | 3 - 4 | 798 | 85 | ||
| XM_005158334.4 | XP_005158391.1 | 5 - 6 | 564 | 85 | ||
| XM_005158335.4 | XP_005158392.1 | 3 - 4 | 568 | 85 | ||
| XM_009292432.3 | XP_009290707.1 | 5 - 6 | 1064 | 85 | ||
| XM_009292434.3 | XP_009290709.1 | 5 - 6 | 564 | 85 | ||
| XM_021466650.1 | XP_021322325.1 | 5 - 6 | 512 | 85 | ||
| GenBank mRNA | BC058052.1 | - | 5 - 6 | 485 | 85 | |
More Information
Set Membership: |
|

phactr4b
Genomic Map