Assay Details
Gene Symbol: paqr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200710.2 | NP_957004.1 | 3 - 4 | 571 | 81 | |
GenBank mRNA | BC059466.1 | - | 3 - 4 | 478 | 81 | |
BC071435.1 | - | 3 - 4 | 571 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
