Assay Details
| Gene Symbol: paqr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200710.2 | NP_957004.1 | 5 - 6 | 926 | 82 | |
| GenBank mRNA | BC059466.1 | - | 5 - 6 | 833 | 82 | |
| BC071435.1 | - | 5 - 6 | 926 | 82 | ||
More Information
Set Membership: |
|

paqr3a
Genomic Map