Assay Details
| Gene Symbol: rps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200752.1 | NP_957046.1 | 4 - 5 | 374 | 77 | |
| GenBank mRNA | AY561508.1 | - | 5 - 6 | 426 | 77 | |
| BC059562.1 | - | 4 - 5 | 374 | 77 | ||
More Information
Set Membership: |
|

rps7
Genomic Map