Assay Details
| Gene Symbol: ppcdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200780.1 | NP_957074.1 | 1 - 2 | 351 | 109 | |
| XM_005159068.4 | XP_005159125.1 | 1 - 2 | 405 | 109 | ||
| XM_009293542.3 | XP_009291817.1 | 1 - 2 | 405 | 109 | ||
| GenBank mRNA | BC059622.1 | - | 1 - 2 | 351 | 109 | |
More Information
Set Membership: |
|

ppcdc
Genomic Map