Assay Details
Gene Symbol: rps17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200845.1 | NP_957139.1 | 2 - 3 | 267 | 109 | |
GenBank mRNA | BC062279.1 | - | 2 - 3 | 267 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
