Assay Details
| Gene Symbol: rab5ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200970.1 | NP_957264.1 | 5 - 6 | 627 | 119 | |
| GenBank mRNA | BC049057.1 | - | 5 - 6 | 627 | 119 | |
More Information
Set Membership: |
|

rab5ab
Genomic Map