Assay Details
Gene Symbol: ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201005.1 | NP_957299.1 | 6 - 7 | 747 | 67 | |
GenBank mRNA | BC047847.1 | - | 6 - 7 | 747 | 67 | |
BC075751.1 | - | 6 - 7 | 711 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
