Assay Details
Gene Symbol: hpda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201167.1 | NP_957461.1 | 5 - 6 | 282 | 119 | |
XM_005171729.3 | XP_005171786.1 | 5 - 6 | 318 | 119 | ||
GenBank mRNA | AY398361.1 | - | 4 - 5 | 246 | 119 | |
BC046075.1 | - | 5 - 6 | 282 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
