Assay Details
Gene Symbol: hpda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201167.1 | NP_957461.1 | 9 - 10 | 638 | 66 | |
XM_005171729.3 | XP_005171786.1 | 9 - 10 | 674 | 66 | ||
GenBank mRNA | AY398361.1 | - | 8 - 9 | 602 | 66 | |
BC046075.1 | - | 9 - 10 | 638 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
