Assay Details
| Gene Symbol: qars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201213.1 | NP_957507.1 | 13 - 14 | 1447 | 84 | |
| GenBank mRNA | BC049420.1 | - | 13 - 14 | 1447 | 84 | |
More Information
Set Membership: |
|

qars1
Genomic Map