Assay Details
| Gene Symbol: fbxw11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201310.1 | NP_958467.1 | 5 - 6 | 660 | 108 | |
| XM_005173069.4 | XP_005173126.1 | 6 - 7 | 799 | 108 | ||
| XM_021480983.1 | XP_021336658.1 | 6 - 7 | 799 | 108 | ||
| GenBank mRNA | BC045356.1 | - | 5 - 6 | 660 | 108 | |
More Information
Set Membership: |
|

fbxw11a
Genomic Map