Assay Details
| Gene Symbol: zgc:73226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205570.2 | NP_991133.2 | 3 - 4 | 350 | 74 | |
| GenBank mRNA | AF493989.1 | - | 3 - 4 | 371 | 74 | |
| BC059568.1 | - | 3 - 4 | 350 | 74 | ||
| BC083199.1 | - | 3 - 4 | 338 | 74 | ||
More Information
Set Membership: |
|

zgc:73226
Genomic Map