Assay Details
| Gene Symbol: zgc:73226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205570.2 | NP_991133.2 | 5 - 6 | 566 | 59 | |
| GenBank mRNA | BC059568.1 | - | 5 - 6 | 566 | 59 | |
| BC083199.1 | - | 5 - 6 | 554 | 59 | ||
More Information
Set Membership: |
|

zgc:73226
Genomic Map