Assay Details
| Gene Symbol: papss2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212562.1 | NP_997727.1 | 9 - 10 | 1269 | 68 | |
| GenBank mRNA | BC047190.1 | - | 9 - 10 | 1269 | 68 | |
| BC068346.1 | - | 9 - 10 | 1250 | 68 | ||
More Information
Set Membership: |
|

papss2b
Genomic Map