Assay Details
Gene Symbol: klf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212746.3 | NP_997911.2 | 2 - 3 | 1384 | 88 | |
GenBank mRNA | AY116305.1 | - | 2 - 3 | 1335 | 88 | |
BC098596.1 | - | 2 - 3 | 1385 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
