Assay Details
| Gene Symbol: rps6ka3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212786.1 | NP_997951.1 | 10 - 11 | 1014 | 69 | |
| XM_005165118.4 | XP_005165175.1 | 10 - 11 | 1229 | 69 | ||
| XM_021476021.1 | XP_021331696.1 | 11 - 12 | 1291 | 69 | ||
| GenBank mRNA | BC045856.1 | - | 10 - 11 | 1014 | 69 | |
| BC164040.1 | - | 10 - 11 | 852 | 69 | ||
More Information
Set Membership: |
|

rps6ka3a
Genomic Map