Assay Details
Gene Symbol: gatad2ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212916.1 | NP_998081.1 | 4 - 5 | 637 | 126 | |
XM_005170444.4 | XP_005170501.1 | 4 - 5 | 928 | 126 | ||
XM_005170445.4 | XP_005170502.1 | 4 - 5 | 928 | 126 | ||
XM_005170446.4 | XP_005170503.1 | 4 - 5 | 929 | 126 | ||
XM_005170447.4 | XP_005170504.2 | 4 - 5 | 658 | 126 | ||
XM_005170448.4 | XP_005170505.1 | 4 - 5 | 682 | 126 | ||
GenBank mRNA | BC067707.1 | - | 4 - 5 | 637 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
