Assay Details
| Gene Symbol: gatad2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212916.1 | NP_998081.1 | 6 - 7 | 825 | 129 | |
| XM_005170445.4 | XP_005170502.1 | 6 - 7 | 1116 | 129 | ||
| GenBank mRNA | BC067707.1 | - | 6 - 7 | 825 | 129 | |
More Information
Set Membership: |
|

gatad2ab
Genomic Map