Assay Details
| Gene Symbol: prkar2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212958.1 | NP_998123.1 | 4 - 5 | 631 | 84 | |
| GenBank mRNA | BC067676.1 | - | 4 - 5 | 631 | 84 | |
More Information
Set Membership: |
|

prkar2ab
Genomic Map