Assay Details
| Gene Symbol: chn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213000.1 | NP_998165.1 | 11 - 12 | 1470 | 64 | |
| GenBank mRNA | AY684586.1 | - | 11 - 12 | 1468 | 64 | |
| BC047837.1 | - | 11 - 12 | 1470 | 64 | ||
| BC083540.1 | - | 5 - 6 | 1017 | 64 | ||
| BC155207.1 | - | 11 - 12 | 1160 | 64 | ||
| BC164679.1 | - | 11 - 12 | 1137 | 64 | ||
| BC165287.1 | - | 5 - 6 | 762 | 64 | ||
More Information
Set Membership: |
|

chn1
Genomic Map