Assay Details
| Gene Symbol: acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213010.2 | NP_998175.2 | 4 - 5 | 363 | 86 | |
| GenBank mRNA | BC045911.1 | - | 4 - 5 | 86 | ||
| BC066533.1 | - | 4 - 5 | 364 | 86 | ||
| BC095591.1 | - | 4 - 5 | 376 | 86 | ||
| BC155272.1 | - | 4 - 5 | 326 | 86 | ||
| BC165798.1 | - | 4 - 5 | 86 | |||
| DT082054.1 | - | 4 - 5 | 303 | 86 | ||
| EB765156.1 | - | 4 - 5 | 363 | 86 | ||
More Information
Set Membership: |
|

acadm
Genomic Map