Assay Details
Gene Symbol: pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213104.1 | NP_998269.1 | 2 - 3 | 585 | 68 | |
GenBank mRNA | BC053415.1 | - | 2 - 3 | 585 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
