Assay Details
| Gene Symbol: ndufs8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213139.1 | NP_998304.1 | 2 - 3 | 84 | 77 | |
| GenBank mRNA | BC058299.1 | - | 2 - 3 | 84 | 77 | |
| BC065459.1 | - | 2 - 3 | 93 | 77 | ||
More Information
Set Membership: |
|

ndufs8a
Genomic Map