Assay Details
| Gene Symbol: csnk1db | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213250.1 | NP_998415.1 | 2 - 3 | 565 | 108 | |
| GenBank mRNA | BC063953.1 | - | 2 - 3 | 565 | 108 | |
| BC097045.1 | - | 2 - 3 | 598 | 108 | ||
| BC165561.1 | - | 2 - 3 | 227 | 108 | ||
| EU127825.1 | - | 2 - 3 | 193 | 108 | ||
More Information
Set Membership: |
|

csnk1db
Genomic Map